Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Carbon disulfide ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

270660-1L 270660-6X1L 270660-100ML 270660-4X2L
BJ270660-1LEA 309.81 USD InStock
BJ270660-1L BJ270660-100ML BJ270660-6X1L BJ270660-4X2L
Carbon disulfide ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Carbon disulphide
  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency


Formula: CS₂
MW: 76.14 g/mol
Boiling Pt: 46 °C (1.013 hPa)
Melting Pt: -76 °C
Density: 1.264 g/cm³ (20 °C)
Flash Pt: -30 °C
Storage Temperature: Ambient
MDL Number: MFCD00011321
CAS Number: 75-15-0
EINECS: 200-843-6
UN: 1131
ADR: 3,I
https://us-legacy.vwr.com/stibo/web/std.lang.all/53/94/785394.jpg
Order Now
Specification Test Results
Identity (IR) complying
Assay (GC) Min. 99.90 %
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.03 %
Absorbance at 750 nm Max. 0.01
Absorbance 500 nm Max. 0.01
Absorbance at 410 nm Max. 0.05
Absorbance at 403 nm Max. 0.10
Absorbance at 394 nm Max. 0.25
Absorbance at 388 nm Max. 0.50
Absorbance at 386 nm Max. 1.0
Autoignition Temperature 212 °F
Explosion Limit 50 %
Vapor Density 2.67 (vs air)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty