Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Disodium hydrogen phosphate dihydrate 98.5-101.0% (dried basis), Puriss. p.a. Reag. Ph. Eur., Fluka™

Supplier: Honeywell Research Chemicals
Synonyms: Sodium hydrogenphosphate dihydrateSorensen's saltdi-Sodium hydrogen orthophosphate dihydrateSodium phosphate dibasic dihydrateSodium monohydrogen phosphate dihydrate

30435-500G 30435-5KG 30435-6X1KG
BJ30435-500GEA 60.14 USD InStock
BJ30435-500G BJ30435-5KG BJ30435-1KG
Disodium hydrogen phosphate dihydrate 98.5-101.0% (dried basis), Puriss. p.a. Reag. Ph. Eur., Fluka™
di-Sodium hydrogen phosphate dihydrate


Formula: Na₂HPO₄·2H₂O
MW: 177.99 g/mol
Melting Pt: 450 °C
Density: 2.07 g/cm³ (16 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149182
CAS Number: 10028-24-7
Order Now
Specification Test Results
Assay (calculated to the dried substance) 98.5 - 101.0 %
Assay of NaH₂PO₄ (calculated to dried substances) Max. 0.5 %
Loss on drying (130°C) 19.5 - 21.0 %
pH (5 %, 20°C) 9.0 - 9.2
Arsenic (As) Max. 0.00005 %
Iron (Fe) Max. 0.0005 %
Potassium (K) Max. 0.005 %
Heavy metals (as Pb) Max. 0.0005 %
Chloride (Cl) Max. 0.001 %
Sulfate (SO₄) Max. 0.005 %
Total N Max. 0.001 %
Appearance of the solution complying
Reducing impurities complying

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty