Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Ammonia solution ∼25% (NH₃ basis) in water, Puriss. p.a. ISO, Reag. Ph. Eur., Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Ammonium hydroxide

30501-6X1L 30501-4X5L 30501-2.5L-GL 30501-2.5L 30501-5L 30501-4X2.5L 30501-6X1L-GL 30501-1L 30501-1L-GL
BJ30501-6X1LCS 508.07 USD InStock
BJ30501-6X1L BJ30501-1L-GL BJ30501-2.5LGL BJ30501-2.5L BJ30501-5L BJ30501-4X2.5L BJ30501-6X1LGL BJ30501-1L BJ30501-4X5L
Ammonia solution ∼25% (NH₃ basis) in water, Puriss. p.a. ISO, Reag. Ph. Eur., Fluka™
Ammonia solution


Formula: NH₃
MW: 35.05 g/mol
Boiling Pt: 37.7 °C (1.013 hPa)
Melting Pt: -92 °C
Density: 0.90 g/cm³ (20 °C)
Flash Pt: 30.2 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS Number: 1336-21-6
UN: 2672
ADR: 8,III
Order Now
Specification Test Results
Assay Min. 25.0 %
Density (D 20/20) 0.892 - 0.910
Non-volatile matter Max. 0.0018 %
Silver (Ag) Max. 0.02 ppm
Aluminium (Al) Max. 0.5 ppm
Gold (Au) Max. 0.1 ppm
Barium (Ba) Max. 0.05 ppm
Bismuth (Bi) Max. 0.1 ppm
Calcium (Ca) Max. 1 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.05 ppm
Chromium (Cr) Max. 0.05 ppm
Copper (Cu) Max. 0.1 ppm
Iron (Fe) Max. 0.1 ppm
Gallium (Ga) Max. 0.02 ppm
Indium (In) Max. 0.02 ppm
Potassium (K) Max. 0.5 ppm
Lithium (Li) Max. 0.02 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.05 ppm
Molybdenum (Mo) Max. 0.05 ppm
Sodium (Na) Max. 1 ppm
Nickel (Ni) Max. 0.05 ppm
Lead (Pb) Max. 0.05 ppm
Platinum (Pt) Max. 0.1 ppm
Tin (Sn) Max. 0.1 ppm
Strontium (Sr) Max. 0.2 ppm
Titanium (Ti) Max. 0.1 ppm
Thallium (Tl) Max. 0.05 ppm
Zinc (Zn) Max. 0.1 ppm
Heavy metals (as Pb) Max. 1 ppm
Carbonate (CO₃) Max. 60 ppm
Carbonate (as CO₂) Max. 10 ppm
Chloride (Cl) Max. 0.5 ppm
Phosphate (PO₄) Max. 0.5 ppm
Silicate (as SiO₂) Max. 10 ppm
Sulfate (SO₄) Max. 2 ppm
Sulfide (S) Max. 0.2 ppm
KMnO₄ reducing matter (as O) Max. 0.0005 %
Readily oxidizable substances complying
Pyridine, related substances (as Pyridine) Max. 2 ppm
Pyridine Max. 0.00005 %
Appearance of the solution complying

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty