Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide solution ≥32% in water for nitrogen analysis (max. 0.0001% N)

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

30531-6X1L 30531-4X5L 30531-4X2.5L
BJ30531-5LCS 912.86 USD InStock
BJ30531-5L BJ30531-2.5L BJ30531-1L
Sodium hydroxide solution ≥32% in water for nitrogen analysis (max. 0.0001% N)
Sodium hydroxide


Formula: NaOH
MW: 40 g/mol
Boiling Pt: 120 °C
Melting Pt: 319 °C
Density: 1.350 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1824
ADR: 8,II
Order Now
Specification Test Results
Assay 31.5 - 32.5 %
Assay of Na₂CO₃ Max. 0.5 %
Aluminium (Al) Max. 0.0005 %
Iron (Fe) Max. 0.0005 %
Heavy metals (as Pb) Max. 0.0005 %
Chloride (Cl) Max. 0.003 %
Phosphate (PO₄) Max. 0.0005 %
Silicate (as SiO₂) Max. 0.001 %
Sulfate (SO₄) Max. 0.0005 %
Total N Max. 0.0001 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty