Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide ≥99.99% (trace metals basis), pellets, Semiconductor grade

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

306576-25G 306576-500G 306576-100G
BJ306576-100GEA 176.27 USD InStock
BJ306576-100G BJ306576-25G BJ306576-500G
Sodium hydroxide ≥99.99% (trace metals basis), pellets, Semiconductor grade
Sodium hydroxide
  • Comprehensive portfolio for the electronics and semiconductor industry
  • For scientists engaged in education, research, development, prototyping and small scale fabrication of high tech micro- and nanochips
  • Superior purity, stability and exceptional lot-to-lot consistency
  • Includes high purity products required for high performance applications such as etchants, solvents or hexamethyldisilazane (HMDS)


Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1.390 °C (1.013 hPa)
Melting Pt: 319 °C
Density: 2.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II
Order Now
Specification Test Results
Vapor Pressure 3 mmHg ( 37 °C)
Appearance of the substance complying
Assay Min. 95.0 %
Assay of Na₂CO₃ Max. 1.0 %
Boron (B) Max. 10 ppm
Cesium (Cs) Max. 20 ppm
Potassium (K) Max. 100 ppm
Lithium (Li) Max. 10 ppm
Rubidium (Rb) Max. 10 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty