Synonyms:
Muriatic acid
30721-6X1L-GL
30721-4X2.5L
30721-2.5L
30721-2.5L-GL
30721-6X1L
BJ307212.5LGLEA
68.76
USD
InStock
BJ307212.5LGL
BJ30721-4X2.5L
BJ30721-1L
BJ30721-2.5L
BJ30721-1L-GL
Hydrochloric acid fuming ≥37%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur. APHA: ≤10, Fluka™
Hydrochloric acid