Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Copper(II) chloride dihydrate ≥99.0% ACS, Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Copper dichloride dihydrate Cupric chloride dihydrate

307483-100G 307483-500G
BJ307483-100GEA 93.25 USD InStock
BJ307483-100G BJ307483-500G
Copper(II) chloride dihydrate ≥99.0% ACS, Fluka™
Copper (II) chloride dihydrate


Formula: CuCl₂·2H₂O
MW: 170.48 g/mol
Boiling Pt: 993 °C (anhydr.)
Melting Pt: 598 °C (anhydr.)
Density: 2.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149674
CAS Number: 10125-13-0
UN: 2802
ADR: 8,III
Order Now
Specification Test Results
Assay 99.0 - 101.0 %
pH (5 %, 20°C) 3.0 - 3.8
Insoluble in hydrochloric acid Max. 0.01 %
Calcium (Ca) Max. 50 ppm
Iron (Fe) Max. 50 ppm
Potassium (K) Max. 100 ppm
Sodium (Na) Max. 200 ppm
Nickel (Ni) Max. 100 ppm
Nitrate (NO₃) Max. 0.015 %
Sulfate (SO₄) Max. 50 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty