Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl acetate ≥99.9%, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Acetic acid ethyl esterAcetic etherAcetidinAcetoxyethaneEthyl acetic esterEthyl ethanoateVinegar naphtha

31063-4X4L 31063-4X2.5L 31063-1L
BJ31063-4LCS 508.03 USD InStock
BJ31063-4L BJ31063-1L BJ31063-2.5L
Ethyl acetate ≥99.9%, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Ethyl acetate
Ethyl Acetate is an organic solvent with the structure CH3-COO-CH2-CH3 (C4H8O2). This colorless liquid has a characteristic sweet odor. In the laboratory, mixtures containing ethyl acetate are used in column chromatography and extractions. Ethyl acetate does not work well as a reaction solvent because it is prone to hydrolysis and transesterification. Ethyl acetate is volatile at room temperature and has a boiling point of 77 °C. Suitable for GC and Pesticide Residue Analysis.

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent


Formula: CH₃COOC₂H₅
MW: 88.11 g/mol
Boiling Pt: 77 °C (1.013 hPa)
Melting Pt: -84 °C
Density: 0.901 g/cm³ (lit.)
Flash Pt: 11.5 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/20/72/722072.jpg
Order Now
Specification Test Results
Autoignition Temperature 801 °F
Assay (GC) Min. 99.9 %
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.02 %
Suitability for residue analysis Max. 5 ng/l
Vapor Density 3 (20 °C, vs air)
Vapor Pressure 73 mmHg ( 20 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty