Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Sodium chloride ≥99.5%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™

Supplier: Honeywell Research Chemicals
Synonyms: HaliteSea saltSaline

31434-6X500G 31434-6X1KG 31434-4X5KG
BJ31434-1KGCS 700.93 USD InStock
BJ31434-1KG BJ31434-5KG BJ31434-500G
Sodium chloride ≥99.5%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™
Sodium chloride


Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1.465 °C (1.013 hPa)
Melting Pt: 800 °C
Density: 2.160 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Order Now
Specification Test Results
Assay Min. 99.5 %
Assay (calculated to the dried substance) 99.0 - 100.5 %
Loss on drying (130°C) Max. 0.2 %
Water insoluble matter Max. 0.005 %
pH (5 %, 25 °C) 5.0 - 8.0
Free acid (as HCl) Max. 0.005 %
Free alkali (as NaOH) Max. 0.005 %
Aluminium (Al) Max. 0.2 ppm
Arsenic (As) Max. 0.5 ppm
Barium (Ba) Max. 0.001 %
Barium (Ba) complying
Calcium (Ca) Max. 0.002 %
Iron (Fe) Max. 1 ppm
Potassium (K) Max. 50 ppm
Magnesium (Mg) Max. 0.0005 %
Mg, earth alkali metals (as Ca) Max. 100 ppm
Heavy metals (as Pb) Max. 3 ppm
Heavy metals (ICP) ACS Max. 5 ppm
Bromide (Br) Max. 50 ppm
Chlorate, Nitrate (as NO₃) Max. 0.003 %
Iodide (I) Max. 0.001 %
Iodide (I) test complying
Hexacyanoferrate(II) ([Fe(CN)₆]₄⁻) Max. 0.0001 %
Ferrocyanides complying
Nitrite (NO₂) complying
Phosphate (PO₄) Max. 5 ppm
Sulfate (SO₄) Max. 10 ppm
Total N Max. 0.001 %
Acidity or alkalinity complying
Appearance of the solution complying

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty