Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium nitrite ≥99%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™

Supplier: Honeywell Research Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger

31443-500G 31443-1KG 31443-250G 31443-100G
BJ31443-250GEA 87.2 USD InStock
BJ31443-250G BJ31443-100G BJ31443-500G BJ31443-1KG
Sodium nitrite ≥99%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™
Sodium nitrite


Formula: NaNO₂
MW: 69 g/mol
Melting Pt: 280 °C
Density: 2.17 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011118
CAS Number: 7632-00-0
EINECS: 231-555-9
UN: 1500
ADR: 5.1,III
Order Now
Specification Test Results
Assay Min. 99.0 %
Water insoluble matter Max. 0.005 %
Calcium (Ca) Max. 0.002 %
Iron (Fe) Max. 0.001 %
Potassium (K) Max. 0.005 %
Heavy metals (as Pb) Max. 0.001 %
Chloride (Cl) Max. 0.005 %
Sulfate (SO₄) Max. 0.005 %

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty