Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium peroxide ≥95%, Puriss. p.a. ACS, ISO, Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: di-Sodium peroxide

31445-100G 31445-6X1KG 31445-500G
BJ31445-100GEA 246.59 USD InStock
BJ31445-100G BJ31445-1KG BJ31445-500G
Sodium peroxide ≥95%, Puriss. p.a. ACS, ISO, Fluka™
Sodium peroxide


Formula: Na₂O₂
MW: 77.98 g/mol
Boiling Pt: Decomposes on heating
Melting Pt: 660 °C
Density: 2.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003497
CAS Number: 1313-60-6
EINECS: 215-209-4
UN: 1504
ADR: 5.1,I
Order Now
Specification Test Results
Assay Min. 95.0 %
Aluminium (Al) Max. 0.001 %
Iron (Fe) Max. 0.002 %
Heavy metals (as Pb) Max. 0.002 %
Chloride (Cl) Max. 0.002 %
Phosphate (PO₄) Max. 5 ppm
Sulfate (SO₄) Max. 0.001 %
Total N Max. 0.003 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty