Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

di-Sodium tetraborate decahydrate ≥99.5%, Puriss. p.a. ACS, ISO buffer substance, Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Sodium tetraborate decahydrateBorax tetra

31457-1KG 31457-100G 31457-5KG 31457-500G
BJ31457-100GEA 78.66 USD InStock
BJ31457-100G BJ31457-500G BJ31457-5KG BJ31457-1KG
di-Sodium tetraborate decahydrate ≥99.5%, Puriss. p.a. ACS, ISO buffer substance, Fluka™
di-Sodium tetraborate decahydrate


Formula: Na₂B₄O₇·10H₂O
MW: 381.37 g/mol
Melting Pt: 742 °C (anhydr.)
Density: 1.7 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149193
CAS Number: 1303-96-4
Order Now
Specification Test Results
Assay 99.5 - 105.0 %
Water insoluble matter Max. 0.001 %
pH (5 %, 20°C) 9.0 - 9.6
pH (0.01 m, 25°C) 9.15 - 9.20
Arsenic (As) Max. 0.0001 %
Calcium (Ca) Max. 0.005 %
Copper (Cu) Max. 0.0005 %
Iron (Fe) Max. 0.0005 %
Lead (Pb) Max. 0.0005 %
Heavy metals (as Pb) Max. 0.001 %
Chloride (Cl) Max. 0.001 %
Phosphate (PO₄) Max. 0.001 %
Sulfate (SO₄) Max. 0.005 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty