Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Silver nitrate ≥99.8%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Silver mononitrateNitric acid silver saltLunar caustic

31630-25G 31630-250G 31630-1KG 31630-100G
BJ31630-250GEA 1641.47 USD InStock
BJ31630-250G BJ31630-1KG BJ31630-25G BJ31630-100G
Silver nitrate ≥99.8%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™
Silver nitrate


Formula: AgNO₃
MW: 169.87 g/mol
Melting Pt: 212 °C
Density: 4.350 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
UN: 1493
ADR: 5.1,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/45/16640645.jpg
Order Now
Specification Test Results
Assay Min. 99.8 %
Acid. or alk. react. impurities complying
Free acid complying
Cadmium (Cd) Max. 0.0001 %
Copper (Cu) Max. 0.0002 %
Iron (Fe) Max. 0.0002 %
Manganese (Mn) Max. 0.0005 %
Lead (Pb) Max. 0.001 %
Zinc (Zn) Max. 0.0001 %
Substances not precipitated by HCl (as SO₄) Max. 0.01 %
Chloride (Cl) Max. 0.0001 %
Nitrite (NO₂) Max. 0.05 %
Sulfate (SO₄) Max. 0.002 %
Al, Pb, Bi and Cu ions complying
Appearance of the solution complying

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty