Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Perdrogen® solution ≥30% (w/w) stabilized, Puriss. p.a. ISO, Reag. Ph. Eur., Fluka™

Supplier: Honeywell Research Chemicals
Danger

31642-1L 31642-5L 31642-500ML 31642-6X1L
BJ31642-6X1LCS 1275.46 USD InStock
BJ31642-6X1L BJ31642-1L BJ31642-5L BJ31642-500ML
Perdrogen® solution ≥30% (w/w) stabilized, Puriss. p.a. ISO, Reag. Ph. Eur., Fluka™
Hydrogen peroxide
Honeywell ACS solvents for general laboratory use are:

  • A value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • Variations and decrease issues traceable to solvent contamination
  • All solvents are tested tmeet ACS general use specifications


Formula: H₂O₂
MW: 34.01 g/mol
Boiling Pt: Decomposes on heating
Melting Pt: -26 °C
Density: 1.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011333
CAS Number: 7722-84-1
UN: 2014
ADR: 5.1,II
Order Now
Specification Test Results
Assay Min. 30 %
Non-volatile matter Max. 0.005 %
Residue on ignition Max. 0.002 %
Free acid (as H₂SO₄) Max. 0.005 %
Acidity complying
Aluminium (Al) Max. 0.00005 %
Arsenic (As) Max. 0.000001 %
Barium (Ba) Max. 0.000005 %
Beryllium (Be) Max. 0.000001 %
Bismuth (Bi) Max. 0.00001 %
Calcium (Ca) Max. 0.00002 %
Cadmium (Cd) Max. 0.000001 %
Cobalt (Co) Max. 0.000001 %
Chromium (Cr) Max. 0.000002 %
Copper (Cu) Max. 0.000001 %
Iron (Fe) Max. 0.000005 %
Germanium (Ge) Max. 0.000005 %
Potassium (K) Max. 0.00001 %
Lithium (Li) Max. 0.000001 %
Magnesium (Mg) Max. 0.000005 %
Manganese (Mn) Max. 0.000001 %
Molybdenum (Mo) Max. 0.000002 %
Sodium (Na) Max. 0.00001 %
Nickel (Ni) Max. 0.000002 %
Lead (Pb) Max. 0.000001 %
Strontium (Sr) Max. 0.000001 %
Titanium (Ti) Max. 0.00001 %
Thallium (Tl) Max. 0.000005 %
Vanadium (V) Max. 0.000001 %
Zinc (Zn) Max. 0.000005 %
Zirconium (Zr) Max. 0.00001 %
Chloride (Cl) Max. 0.00005 %
Phosphate (PO₄) Max. 0.0001 %
Sulfate (SO₄) Max. 0.0001 %
Total N Max. 0.0002 %
Organic stabilisators Max. 0.05 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty