Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Diethyl ether ≥99.8% stabilized, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: 2,2'-Oxybisethane2,2'-OxydiethaneEtherEthoxyethaneEthyl ether

Stabilization: Stabilised with ethanol 2%

31671-4X4L 31671-6X1L
BJ31671-4LCS 1001.89 USD InStock
BJ31671-4L BJ31671-1L
Diethyl ether ≥99.8% stabilized, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Diethyl ether


Formula: (CH₃CH₂)₂O
MW: 74.12 g/mol
Boiling Pt: 35 °C
Melting Pt: -117.4 °C
Density: 0.713 g/cm³ (20 °C)
Flash Pt: -45 °C
Storage Temperature: Ambient
MDL Number: MFCD00011646
CAS Number: 60-29-7
EINECS: 200-467-2
UN: 1155
ADR: 3,I
Order Now
Specification Test Results
Assay GC (Product + Ethanol) Min. 99.8 %
Refractive Index n20/D 1.353
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.05 %
Autoignition Temperature 320 °F
Refractive index (n 20/D) 1.352 - 1.354
Suitability for residue analysis Max. 5 ng/l
Explosion Limit 36.5 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty