Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 0.5 M HCl (0.5N) volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Warning
Synonyms: Muriatic acidHydrochloric acid solution

318957-500ML 318957-2L
BJ318957-500MLEA 46.19 USD InStock
BJ318957-500ML BJ318957-2L
Hydrochloric acid 0.5 M HCl (0.5N) volumetric solution, Fluka™
Hydrochloric acid


Formula: HCl
MW: 36.46 g/mol
Boiling Pt: >100 °C (1.013 hPa)
Melting Pt: 0 °C
Density: 1.010 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8,III
Order Now
Specification Test Results
Assay 0.4900 - 0.5100 N
Traceable to NIST SRM effective

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty