Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Silver nitrate 1.0 M AgNO3 (1.0N) volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: Silver mononitrateNitric acid silver saltLunar caustic

319449-500ML 319449-1L
BJ319449-1LEA 734.28 USD InStock
BJ319449-1L BJ319449-500ML
Silver nitrate 1.0 M AgNO3 (1.0N) volumetric solution, Fluka™
Silver nitrate


Formula: AgNO₃
MW: 169.87 g/mol
Boiling Pt: 100 °C
Density: 1.140 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
UN: 3264
ADR: 8,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/45/16640645.jpg
Order Now
Specification Test Results
Appearance complying
Coloration complying
Clearness complying
Assay titr. 0.950 - 1.050 mol/l
Traceable to NIST SRM complying
BAM/EMPA reference material complying
BAM/EMPA Certificate Number complying

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty