Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide 0.1 M NaOH (0.1N) ≥99% volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Warning
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

319481-500ML 319481-6X500ML 319481-2L 319481-1L 319481-2.5L
BJ319481-2.5LEA 154.4 USD InStock
BJ319481-2.5L BJ319481-500ML BJ319481-1L BJ319481-2L BJ3194816500ML
Sodium hydroxide 0.1 M NaOH (0.1N) ≥99% volumetric solution, Fluka™
Sodium hydroxide


Formula: NaOH
MW: 40 g/mol
Boiling Pt: 100 °C (1.013 hPa)
Melting Pt: 319 °C
Density: 1.000 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 231-791-2
UN: 1824
ADR: 8,III
Order Now
Specification Test Results
Assay titr. 0.095 - 0.105 mol/l
Carbonate (CO₃) Max. 0.0015 mol/l
Traceable to NIST SRM effective
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty