Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Ethyl acetate ≥99.5% ACS, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: EtOAcAcetidinEthyl ethanoateVinegar naphthaEthyl acetic esterAcetic acid ethyl esterAcetic etherAcetoxyethane

319902-6X500ML 319902-6X1L 319902-500ML
BJ3199026500MLCS 199.53 USD InStock
BJ3199026500ML BJ319902-6X1L BJ319902-500ML
Ethyl acetate ≥99.5% ACS, Riedel-de Haën™
Ethyl acetate
ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Beilstein Number: 506104
RTECS Number: AH5425000
UNSPSC Code: 12191602


Formula: CH₃COOC₂H₅
MW: 88.11 g/mol
Boiling Pt: 77 °C (1.013 hPa)
Melting Pt: -84 °C
Density: 0.901 g/cm³
Flash Pt: 11.5 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/20/72/722072.jpg
Order Now
Specification Test Results
Identity (IR) complying
Assay (GC) Min. 99.50 %
Non-volatile matter Max. 0.003 %
Water (Karl Fischer) Max. 0.2 %
Titrable Acid Max. 0.0009 meq/g
Reaction against H₂SO₄ complying
APHA Max. 10
Vapor Pressure 73 mmHg ( 20 °C)
Autoignition Temperature 801 °F
Vapor Density 3 (20 °C, vs air)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty