Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium bromide concentrate for dilution to 1L, 0.1 M KBr volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals

32036-1EA
BJ32036-1EAEA 94.79 USD InStock
BJ32036-1EA
Potassium bromide concentrate for dilution to 1L, 0.1 M KBr volumetric solution, Fluka™
Potassium bromide


Formula: KBr
MW: 119 g/mol
Density: 1.0 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011358
CAS Number: 7758-02-3
Order Now
Specification Test Results
Factor 0.998 - 1.002

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty