Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Acetone ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Dimethyl ketoneDimethylformaldehydeDMKPropanone2-Propanone

32201-4X2.5L 32201-1L 32201-2.5L-GL 32201-20KG 32201-6X1L 32201-25L 32201-2.5L 32201-4X2.5L-GL
BJ322012.5LGLEA 119.21 USD InStock
BJ322012.5LGL BJ32201-6X1L BJ32201-1L BJ32201-20KG BJ32201-2.5L BJ3220142.5LGL BJ32201-25L BJ32201-4X2.5L
Acetone ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Acetone
ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Beilstein Number: 635680
RTECS Number: AL3150000
UNSPSC Code: 12191602


Formula: CH₃COCH₃
MW: 58.08 g/mol
Boiling Pt: 55-57 °C (1.013 hPa)
Melting Pt: -95 °C
Density: 0.79 g/cm³ (20 °C)
Flash Pt: 14.3 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/07/720607.jpg
Order Now
Specification Test Results
Assay (GC) Min. 99.5 %
Boiling range 55.5 - 56.5 °C
Density (D 20/4) 0.790 - 0.792
Refractive index (n 20/D) 1.3580 - 1.3600
Non-volatile matter Max. 10 ppm
Water insoluble substances complying
Water (Karl Fischer) Max. 0.2 %
Free acid (as CH₃COOH) Max. 0.0015 %
Free alkali (as NH₃) Max. 0.0008 %
Acidity or alkalinity complying
Aluminium (Al) Max. 0.5 ppm
Boron (B) Max. 0.02 ppm
Barium (Ba) Max. 0.1 ppm
Bismuth (Bi) Max. 0.1 ppm
Calcium (Ca) Max. 0.5 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.01 ppm
Chromium (Cr) Max. 0.02 ppm
Copper (Cu) Max. 0.02 ppm
Iron (Fe) Max. 0.1 ppm
Potassium (K) Max. 0.5 ppm
Lithium (Li) Max. 0.1 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.01 ppm
Molybdenum (Mo) Max. 0.1 ppm
Sodium (Na) Max. 0.5 ppm
Nickel (Ni) Max. 0.02 ppm
Lead (Pb) Max. 0.02 ppm
Tin (Sn) Max. 0.1 ppm
Strontium (Sr) Max. 0.1 ppm
Zinc (Zn) Max. 0.05 ppm
KMnO₄ reducing matter (as O) Max. 0.0002 %
Reducing impurities complying
Aldehydes (as HCHO) Max. 0.001 %
Benzene (GC) Max. 2 Vol.ppm
Ethanol (GC) Max. 0.01 %
Methanol (GC) Max. 0.05 Vol.%
2-propanol (GC) Max. 0.05 Vol.%
Related substances (GC) complying
Appearance of the solution complying
APHA Max. 10
Solubility in water complying

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty