Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Diethyl ether ≥99.8% contains inhibitor, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Ether2,2'-OxybisethaneEthoxyethaneEthyl Ether2,2'-OxydiethaneEthyl ether

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

32203-4X2.5L 32203-2.5L 32203-4X5L 32203-6X1L 32203-27L 32203-1L 32203-5L
BJ32203-1LEA 133.49 USD InStock
BJ32203-1L BJ32203-2.5L BJ32203-6X1L BJ32203-27L BJ32203-4X5L BJ32203-4X2.5L BJ32203-5L
Diethyl ether ≥99.8% contains inhibitor, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Diethyl ether
ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Beilstein Number: 1696894
RTECS Number: KI5775000
UNSPSC Code: 12191602


Formula: (CH₃CH₂)₂O
MW: 74.12 g/mol
Boiling Pt: 34-35 °C (1.013 hPa)
Melting Pt: -116 °C (1.013 hPa)
Density: 0.71 g/cm³ (20 °C)
Flash Pt: -45 °C (1.013 hPa)
Storage Temperature: Ambient
MDL Number: MFCD00011646
CAS Number: 60-29-7
EINECS: 200-467-2
UN: 1155
ADR: 3,I
https://us-legacy.vwr.com/stibo/web/std.lang.all/02/58/730258.jpg
Order Now
Specification Test Results
Appearance (color) complying
Appearance (form) complying
Assay (GC) Min. 99.8 %
Boiling range 34 - 35 °C
Density (D 20/4) 0.713 - 0.714
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.03 %
Free acid (as CH₃COOH) Max. 0.0002 %
Titrable Acid Max. 0.0002 meq/g
Aluminium (Al) Max. 0.50 ppm
Boron (B) Max. 0.02 ppm
Barium (Ba) Max. 0.10 ppm
Calcium (Ca) Max. 0.50 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.02 ppm
Chromium (Cr) Max. 0.02 ppm
Copper (Cu) Max. 0.02 ppm
Iron (Fe) Max. 0.05 ppm
Magnesium (Mg) Max. 0.10 ppm
Manganese (Mn) Max. 0.02 ppm
Nickel (Ni) Max. 0.02 ppm
Lead (Pb) Max. 0.05 ppm
Tin (Sn) Max. 0.10 ppm
Zinc (Zn) Max. 0.05 ppm
Acetone (GC) Max. 0.005 %
Aldehydes complying
Carbonyl compounds (as CO) Max. 0.001 %
Carbonyl Compounds (as HCHO) Max. 0.001 %
Ethanol (GC) Max. 0.02 %
Methanol (GC) Max. 0.02 %
Peroxides (as H₂O₂) Max. 0.3 ppm
Reaction against H₂SO₄ complying
APHA Max. 10
Assay BHT (GC) 5 - 8 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty