Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Methanol ≥99.8% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: CarbinolMethyl alcoholMethyl hydroxideMethylolWood alcohol

32213-2.5L 32213-4X2.5L 32213-1L 32213-4X5L 32213-150KG 32213-6X1L 32213-5L 32213-1L-GL 32213-25L 32213-45KG
BJ32213-5LEA 183.5 USD InStock
BJ32213-5L BJ32213-45KG BJ32213-6X1L BJ32213-4X5L BJ32213-1L-GL BJ32213-4X2.5L BJ32213-150KG BJ32213-1L BJ32213-2.5L BJ32213-25L
Methanol ≥99.8% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Methanol
ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Beilstein Number: 1098229


Formula: CH₃OH
MW: 32.04 g/mol
Boiling Pt: 64-65 °C (1.013 hPa)
Melting Pt: -98 °C
Density: 0.79 g/cm³ (20 °C)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/33/12/733312.jpg
Order Now
Specification Test Results
Appearance (form) complying
Appearance (color) complying
Appearance complying
Assay (GC) Min. 99.8 %
Boiling range 64 - 65 °C
Density (D 20/20) 0.791 - 0.793
Refractive index (n 20/D) 1.328 - 1.330
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.05 %
Free acid (as HCOOH) Max. 0.002 %
Free alkali (as NH₃) Max. 0.00005 %
Aluminium (Al) Max. 0.00005 %
Boron (B) Max. 0.000002 %
Barium (Ba) Max. 0.00001 %
Bismuth (Bi) Max. 0.00001 %
Calcium (Ca) Max. 0.00005 %
Cadmium (Cd) Max. 0.000005 %
Cobalt (Co) Max. 0.000002 %
Chromium (Cr) Max. 0.000002 %
Copper (Cu) Max. 0.000001 %
Iron (Fe) Max. 0.00001 %
Potassium (K) Max. 0.00005 %
Lithium (Li) Max. 0.00001 %
Magnesium (Mg) Max. 0.00001 %
Manganese (Mn) Max. 0.000001 %
Molybdenum (Mo) Max. 0.00001 %
Sodium (Na) Max. 0.00005 %
Nickel (Ni) Max. 0.000002 %
Lead (Pb) Max. 0.000002 %
Tin (Sn) Max. 0.00001 %
Strontium (Sr) Max. 0.00001 %
Zinc (Zn) Max. 0.00001 %
Chloride (Cl) Max. 0.00005 %
Sulfate (SO₄) Max. 0.0001 %
KMnO₄ reducing matter (as O) Max. 0.00025 %
Acetone (GC) Max. 0.001 %
Ketones (as CH₃COCH₃) Max. 0.001 %
Aldehydes (as CH₃CHO) Max. 0.001 %
Acetaldehyde Max. 0.001 %
Ethanol (GC) Max. 0.1 %
Formaldehyde Max. 0.001 %
Carbonyl compounds (as CO) Max. 0.005 %
Carbonyl compounds (ACS) complying
Mixable with H₂O complying
Reaction against H₂SO₄ complying
APHA Max. 10

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty