Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrogen peroxide 3% (w/w) in water stabilized, Fluka™

Supplier: Honeywell Research Chemicals
Danger

Stabilization: Stabilised with acetanilide ~200 ppm

323381-4L 323381-500ML 323381-25ML
BJ323381-500MLEA 90.92 USD InStock
BJ323381-500ML BJ323381-4L BJ323381-25ML
Hydrogen peroxide 3% (w/w) in water stabilized, Fluka™
Hydrogen peroxide
contains ∼200 ppm acetanilide as stabilizer


Formula: H₂O₂
MW: 34.01 g/mol
Melting Pt: –26 °C
Density: 1.00 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011333
CAS Number: 7722-84-1
ADR: 5.1,II
Order Now
Specification Test Results
Appearance (colorless liquid) complying
Assay 3.0 - 4.0 Gew.%
pH (direct, 20 °C) 4 - 7
Stabilizer (Acetanilide) 150 - 250 ppm
Contains ~200 ppm acetanilide as stabilizer

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty