Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Acetic acid ≥99.8%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Methanecarboxylic acidVinegar acidEthanoic acid

33209-6X1L-GL 33209-2.5L-GL 33209-60KG 33209-4X2.5L 33209-30KG 33209-6X1L 33209-5L
BJ33209-5LEA 296.51 USD InStock
BJ33209-5L BJ33209-1L-GL BJ33209-1L BJ33209-60KG BJ33209-2.5L BJ33209-30KG BJ33209-2.5LGL
Acetic acid ≥99.8%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™
Acetic acid


Formula: H₃CCOOH
MW: 60.05 g/mol
Boiling Pt: 118 °C (1.013 hPa)
Melting Pt: 16 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 40 °C
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8,II
Order Now
Specification Test Results
Assay 99.8 - 100.5 %
Assay (GC) Min. 99.8 %
Density (D 20/20) 1.052 - 1.053
Boiling range 117 - 119 °C
Congealing - Freezing Point Min. 16.3 °C
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.4 %
Silver (Ag) Max. 0.000001 %
Aluminium (Al) Max. 0.000005 %
Arsenic (As) Max. 0.000001 %
Barium (Ba) Max. 0.000001 %
Beryllium (Be) Max. 0.000001 %
Bismuth (Bi) Max. 0.00001 %
Calcium (Ca) Max. 0.00002 %
Cadmium (Cd) Max. 0.000002 %
Cobalt (Co) Max. 0.000001 %
Chromium (Cr) Max. 0.000005 %
Copper (Cu) Max. 0.000001 %
Iron (Fe) Max. 0.00002 %
Germanium (Ge) Max. 0.000005 %
Potassium (K) Max. 0.00001 %
Lithium (Li) Max. 0.000001 %
Magnesium (Mg) Max. 0.00001 %
Manganese (Mn) Max. 0.000001 %
Molybdenum (Mo) Max. 0.000002 %
Sodium (Na) Max. 0.00005 %
Nickel (Ni) Max. 0.000005 %
Lead (Pb) Max. 0.000002 %
Strontium (Sr) Max. 0.000001 %
Titanium (Ti) Max. 0.000001 %
Thallium (Tl) Max. 0.000005 %
Vanadium (V) Max. 0.000001 %
Zinc (Zn) Max. 0.000005 %
Zirconium (Zr) Max. 0.00001 %
Heavy metals (as Pb) Max. 0.00005 %
Chloride (Cl) Max. 0.00005 %
Phosphate (PO₄) Max. 0.00005 %
Sulfate (SO₄) Max. 0.00005 %
Acetaldehyde Max. 0.0002 %
Acetic aNHydride (GC) Max. 0.01 %
Mixable with H₂O complying
Titrable Base Max. 0.0004 meq/g
APHA Max. 10
Appearance of the substance complying
K₂Cr₂O₇ reducing matter complying
KMnO₄ reducing substances complying

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty