Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Acetic acid ethyl esterAcetic etherAcetidinAcetoxyethaneEthyl acetic esterEthyl ethanoateVinegar naphtha

33211-180KG 33211-2.5L 33211-4X2.5L 33211-1L 33211-6X1L 33211-25L
BJ33211-180KGEA 5151.56 USD InStock
BJ33211-180KG BJ33211-2.5L BJ33211-1L BJ33211-6X1L BJ33211-4X2.5L BJ33211-25L
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Ethyl acetate
ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Beilstein Number: 506104
RTECS Number: AH5425000
UNSPSC Code: 12191602


Formula: CH₃COOC₂H₅
MW: 88.11 g/mol
Boiling Pt: 77 °C (1.013 hPa)
Melting Pt: -84 °C
Density: 0.901 g/cm³
Flash Pt: 11.5 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/20/72/722072.jpg
Order Now
Specification Test Results
Assay (GC) Min. 99.5 %
Boiling range 76 - 78 °C
Density (D 20/4) 0.899 - 0.901
Refractive index (n 20/D) 1.3710 - 1.3730
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.05 %
Free acid (as CH₃COOH) Max. 0.0045 %
Aluminium (Al) Max. 0.00005 %
Boron (B) Max. 0.000002 %
Barium (Ba) Max. 0.00001 %
Bismuth (Bi) Max. 0.00001 %
Calcium (Ca) Max. 0.00005 %
Cadmium (Cd) Max. 0.000005 %
Cobalt (Co) Max. 0.000002 %
Chromium (Cr) Max. 0.000002 %
Copper (Cu) Max. 0.000002 %
Iron (Fe) Max. 0.00001 %
Potassium (K) Max. 0.00005 %
Lithium (Li) Max. 0.00001 %
Magnesium (Mg) Max. 0.00001 %
Molybdenum (Mo) Max. 0.00001 %
Manganese (Mn) Max. 0.000002 %
Sodium (Na) Max. 0.00005 %
Nickel (Ni) Max. 0.000002 %
Lead (Pb) Max. 0.00001 %
Tin (Sn) Max. 0.00001 %
Strontium (Sr) Max. 0.00001 %
Zinc (Zn) Max. 0.00001 %
methyl acetate Max. 0.1 %
Ethanol (GC) Max. 0.1 %
Isobutyl alcohol (GC) Max. 0.1 %
Methanol (GC) Max. 0.1 %
Reaction against H₂SO₄ complying
APHA Max. 10

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty