Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Murexide for complexometry, Fluka™

Supplier: Honeywell Research Chemicals
Synonyms: Murexide triturateAmmonium 5-(2,4,6-trioxoperhydropyrimidin-5-ylideneamino)barbituratePurpuric acid ammonium saltAmmonium purpurate

33414-25G 33414-10G 33414-5G
BJ33414-25GEA 205.84 USD InStock
BJ33414-25G BJ33414-5G BJ33414-10G
Murexide for complexometry, Fluka™
Murexide


Formula: C₈H₈N₆O₆
MW: 284.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012777
CAS Number: 3051-09-0
EINECS: 221-266-6
Order Now
Specification Test Results
Suitability for Complexometry complying

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty