Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Molecular sieve 3A (0.3 nm, 3 Å), pellets, Fluka™

Supplier: Honeywell Research Chemicals
Synonyms: Linde 3AMolecular sieve A33A molecular sieve

334278-1KG
BJ334278-1KGEA 249.72 USD InStock
BJ334278-1KG
Molecular sieve 3A (0.3 nm, 3 Å), pellets, Fluka™
Molecular sieve 3A (0.3 nm, 3 Å)


Formula: KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O
Melting Pt: 400 °C
Density: 2 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00131613
CAS Number: 308080-99-1
Order Now
Specification Test Results
Particle Size 3.2 mm
Form pellets
Granulation complying
Pore Size (Ång Ström) 3

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty