Would you like to log in again?
pH Buffer Solutions, Referenced at 20 °C, Traceable to NIST, Fluka™, Honeywell Research Chemicals
Supplier: Honeywell Research Chemicals
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
33643-100ML 33648-5L-VP 31046-6X1L 33648-6X1L 31044-1L 33646-5L-VP 31045-100ML 33643-10L-VP 33646-10L-VP 31045-1L 33643-6X1L 33544-6X1L 33643-5L-VP 33646-6X1L 33547-6X1L 33545-6X1L 33648-10L-VP 33643-500ML
BJ33643-100MLEA 37 USD InStock
BJ33643-100ML BJ33547-1L BJ33648-5L-VP BJ31045-100ML BJ33643-10L-VP BJ33643-5L-VP BJ33544-1L BJ33545-1L BJ33643-1L BJ33648-10L-VP BJ33646-5L-VP BJ33646-1L BJ33646-10L-VP BJ33648-1L BJ31044-1L BJ33643-500ML BJ31046-1L BJ31045-1L
pH Buffer Solutions, Referenced at 20 °C, Traceable to NIST, Fluka™, Honeywell Research Chemicals
Buffers pH Calibration Buffers
  • Citrate and phosphate buffer solutions contain fungicide
  • Traceable to SRM from NIST
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty