Would you like to log in again?
HYDRANAL™ - NEXTGEN solvents for Karl Fischer titration, Fluka™
Supplier: Honeywell Research Chemicals
34432-6X1L 34431-6X1L 34431-1L 34432-1L
BJ34432-6X1LBX 645.91 USD InStock
BJ34432-6X1L BJ34431-1L BJ34432-1L BJ34431-6X1L
HYDRANAL™ - NEXTGEN solvents for Karl Fischer titration, Fluka™
Karl Fischer Titrants and Reagents
Medium for volumetric two-component Karl Fischer titration (ethanol or methanol based), Free of imidazole (free of CMR substances)
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty