Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zinc sulfate 0.05 M ZnSO4 ≥99% volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Warning

34448-1L
BJ34448-1LEA 221.25 USD InStock
BJ34448-1L
Zinc sulfate 0.05 M ZnSO4 ≥99% volumetric solution, Fluka™
Zinc sulphate heptahydrate


Formula: ZnSO₄·7H₂O
MW: 287.56 g/mol
Boiling Pt: 100 °C (solvent)
Melting Pt: <0 °C
Density: 1.005 g/cm³
Flash Pt: Does not flash
Storage Temperature: Ambient
MDL Number: MFCD00149894
CAS Number: 7446-20-0
Order Now
Specification Test Results
Factor 0.998 - 1.002

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty