Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

n-Heptane ≥99% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: DipropylmethaneEptaniHeptanHeptaneHeptanenHeptyl hydrideSkellysolve c

34495-4X4L 34495-1L 34495-4X2.5L
BJ34495-4LCS 1452.57 USD InStock
BJ34495-4L BJ34495-2.5L BJ34495-1L
n-Heptane ≥99% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
n-Heptane
n-Heptane is the straight-chain alkane with the chemical formula H3C(CH2)5CH3 or C7H16. Heptane is a Greener alternative to Hexane in the laboratory, with lower long term toxicity. Suitable for GC and Pesticide Residue Analysis.

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent


Formula: H₃C(CH₂)₅CH₃
MW: 100.20 g/mol
Boiling Pt: 98.2 - 98.4 °C (lit.)
Melting Pt: -90.5 °C (lit.)
Density: 0.69 g/cm³ (15 °C) (lit.)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009544
CAS Number: 142-82-5
EINECS: 205-563-8
UN: 1206
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/64/15/736415.jpg
Order Now
Specification Test Results
Assay (GC) Min. 99 %
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.01 %
Suitability for residue analysis Max. 5 ng/l
Autoignition Temperature 433 °F
Refractive Index n20/D 1.387 (lit.)
Determination of P/N ≤5 ng/L
Vapor Pressure 83 mmHg ( 37.7 °C)
Explosion Limit 7 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty