Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

n-Pentane ≥99% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™

Supplier: Honeywell Research Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: Pentane

34497-6X1L 34497-2.5L
BJ34497-2.5LEA 360.51 USD InStock
BJ34497-2.5L BJ34497-1L
n-Pentane ≥99% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
n-Pentane


Formula: H₃C(CH₂)₃CH₃
MW: 72.15 g/mol
Boiling Pt: 36-37 °C
Melting Pt: -131 °C
Density: 0.631 g/cm³ (15 °C) (lit.)
Flash Pt: <-35 °C
Storage Temperature: Ambient
MDL Number: MFCD00009498
CAS Number: 109-66-0
EINECS: 203-692-4
UN: 1265
ADR: 3,II
Order Now
Specification Test Results
Assay (GC) Min. 99 %
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.01 %
Suitability for residue analysis Max. 5 ng/l
Explosion Limit ~8.3 %
Autoignition Temperature 500 °F

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty