Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acetonitrile ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: CyanomethaneEthanenitrileEthanoic acid nitrileMethane, cyano-Methyl cyanideMethanecarbonitrileACN

34851-200L 34851-4X2.5L 34851-4X2.5L 34851-1L 34851-4X4L 34851-56L
BJ34851-4LCS 1870.05 USD InStock
BJ34851-4L BJ34851-1L BJ34851-4X2.5L BJ34851-200L BJ34851-56L BJ34851-2.5L
Acetonitrile ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
Acetonitrile
  • Same high quality as CHROMASOLV HPLC
  • Additional testing for HPLC via gradient analysis and UV-VIS spectroscopy
  • Low UV profile by HPLC
  • Low impurity gradient LC baseline

Beilstein Number: 741857


Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 82 °C (1.013 hPa)
Melting Pt: -46 °C
Density: 0.7857 g/cm³ (20 °C) (lit.)
Flash Pt: 6 °C
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/89/10/718910.jpg
Order Now
Specification Test Results
Assay (GC) Min. 99.90 %
Refractive index (n 20/D) 1.3435 - 1.3445
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.02 %
Free acid (as CH₃COOH) Max. 0.001 %
Free alkali (as NH₃) Max. 0.0002 %
Absorbance at 400 nm Max. 0.0044
Absorbance at 250 nm Max. 0.0044
Absorbance at 235 nm Max. 0.0044
Absorbance at 230 nm Max. 0.0044
Absorbance at 200 nm Max. 0.032
Absorbance at 195 nm Max. 0.12
Fluorescence (chinin) at 254 nm Max. 1 ppb
Fluorescence (chinin) at 365 nm Max. 0.5 ppb
HPLC Gradient at 210 nm Max. 3 mAU
HPLC Gradient at 254 nm Max. 0.5 mAU
Baseline drift at 210 nm Max. 15 mAU
Refractive Index n20/D 1.344 (lit.)
Measuring Range ≤15 mAU baseline drift, (210 nm)
Autoignition Temperature 973 °F

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty