Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Dichloromethane ≥99.8% stabilized, CHROMASOLV™ for HPLC, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Warning
Synonyms: Methane dichlorideMethylene chloride

Stabilization: Stabilised with amylene

34856-2.5L 34856-4X4L 34856-4X2.5L 34856-1L
BJ34856-4X2.5LCS 602.43 USD InStock
BJ34856-4X2.5L BJ34856-1L BJ34856-2.5L BJ34856-4L
Dichloromethane ≥99.8% stabilized, CHROMASOLV™ for HPLC, Riedel-de Haën™
Dichloromethane
Dichloromethane (DCM, Methylene chloride) is an organic compound with formula CH2Cl2. This colorless, volatile liquid has a moderately sweet odor and is widely used as a solvent. Although it is not miscible with water, it is miscible with many organic solvents. Suitable for HPLC and spectrophotometry.

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency


Formula: CH₂Cl₂
MW: 84.93 g/mol
Boiling Pt: 39-41 °C (1.013 hPa)
Melting Pt: -97 °C
Density: 1.32-1.33 g/cm³ (20 °C) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/14/30/20961430.jpg
Order Now
Specification Test Results
Appearance complying
Identity (IR) complying
Assay (GC) Min. 99.80 %
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.02 %
Free acid (as HCl) Max. 0.001 %
Free chlorine complying
Absorbance at 260 nm Max. 0.01
Absorbance at 245 nm Max. 0.10
Absorbance at 240 nm Max. 0.30
Absorbance at 235 nm Max. 0.70
Absorbance complying
APHA Max. 10
2-methyl-2-buten (GC) 10 - 60 ppm
Autoignition Temperature 1223 °F
Contains amylene as stabilizer
Refractive Index n20/D 1.424 (lit.)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty