Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl ethyl ketone ≥99.7%, CHROMASOLV™ for HPLC, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: 2-ButanoneMEKButan-2-oneEK 500 AdditiveButanoneEthyl methyl ketone

34861-1L 34861-4X2.5L 34861-2.5L 34861-6X1L 34861-100ML 34861-4X4L
BJ34861-6X1LCS 740.61 USD InStock
BJ34861-6X1L BJ34861-100ML BJ34861-2.5L BJ34861-1L BJ34861-4X2.5L BJ34861-4X4L
Methyl ethyl ketone ≥99.7%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Methyl ethyl ketone
  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency

Beilstein Number: 741880


Formula: C₂H₅COCH₃
MW: 72.11 g/mol
Boiling Pt: 79-81 °C
Melting Pt: -87 °C
Density: 0.805 g/cm³ (20 °C)
Flash Pt: 11.5 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00011648
CAS Number: 78-93-3
UN: 1193
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/03/79/720379.jpg
Order Now
Specification Test Results
Assay (GC) Min. 99.7 %
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.1 %
Free acid (as CH₃COOH) Max. 0.001 %
Transmittance at 335 nm Min. 50 %
Transmittance at 340 nm Min. 80 %
Transmittance from 350 nm Min. 98 %
Explosion Limit 10.1 %
Autoignition Temperature 960 °F

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty