Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Toluene ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Benzene, methylMethacideMethane, phenyl-MethylbenzeneMethylbenzolPhenylmethaneToluol

34866-100ML 34866-2.5L 34866-6X1L 34866-4X4L 34866-4X2.5L
BJ34866-1LCS 504.33 USD InStock
BJ34866-1L BJ34866-100ML BJ34866-2.5L BJ34866-4X2.5L BJ34866-4L
Toluene ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Toluene


Formula: C₆H₅CH₃
MW: 92.14 g/mol
Boiling Pt: 110.6 °C
Melting Pt: -95 °C
Density: 0.867 g/cm³ (20 °C) (lit.)
Flash Pt: 7.1 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3,II
Order Now
Specification Test Results
Assay (GC) Min. 99.9 %
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.02 %
Free acid (as HCl) Max. 0.001 %
Thiophene Max. 0.0005 %
Absorbance at 400 nm Max. 0.01
Absorbance at 350 nm Max. 0.01
Absorbance at 300 nm Max. 0.10
Absorbance at 288 nm Max. 0.50
Absorbance at 286 nm Max. 1.00
Refractive Index n20/D 1.496 (lit.)
Explosion Limit 7 %
Vapor Pressure 22 mmHg ( 20 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty