Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acetonitrile ≥99.8% (by GC), CHROMASOLV™ for liquid chromatography, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: CyanomethaneEthanenitrileEthanoic acid nitrileMethane, cyano-Methyl cyanideMethanecarbonitrileACN

34881-6X1L 34881-4X2.5L 34881-4X4L
BJ34881-1LCS 906.84 USD InStock
BJ34881-1L BJ34881-2.5L BJ34881-4L
Acetonitrile ≥99.8% (by GC), CHROMASOLV™ for liquid chromatography, Riedel-de Haën™
Acetonitrile
Acetonitrile is a medium-polarity solvent that is miscible with water and a range of organic solvents, but not saturated hydrocarbons. CHROMASOLV HPLC-GC Acetonitrile is suitable for multiple applications including: HPLC and GC applications.


Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 82 °C (1.013 hPa)
Melting Pt: -46 °C
Density: 0.7857 g/cm³ (20 °C)
Flash Pt: 6 °C
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/89/10/718910.jpg
Order Now
Specification Test Results
Assay (GC) min. 99.8 %
Non-volatile matter max. 0.0005 %
Water (Karl Fischer) max. 0.1 %
Free acid (as CH₃COOH) max. 0.001 %
Transmittance at 200 nm min. 50 %
Transmittance at 220 nm min. 90 %
Transmittance from 240 nm min. 99 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty