Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

n-Pentane ≥99.0%, CHROMASOLV™ for HPLC, Riedel-de Haën™

Supplier: Honeywell Research Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: Pentane

34956-2.5L 34956-4X4L 34956-1L 34956-6X1L 34956-4X2.5L
BJ34956-1LEA 186.45 USD InStock
BJ34956-1L BJ34956-2.5L BJ34956-4X2.5L BJ34956-4X4L BJ34956-6X1L
n-Pentane ≥99.0%, CHROMASOLV™ for HPLC, Riedel-de Haën™
n-Pentane
  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency

Beilstein Number: 969132


Formula: H₃C(CH₂)₃CH₃
MW: 72.15 g/mol
Boiling Pt: 36-37 °C
Melting Pt: -131 °C
Density: 0.631 g/cm³ (15 °C)
Flash Pt: <-35 °C
Storage Temperature: Ambient
MDL Number: MFCD00009498
CAS Number: 109-66-0
UN: 1265
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/81/67/738167.jpg
Order Now
Specification Test Results
Assay (GC) Min. 99.0 %
IR spectrum complying
Refractive index (n 20/D) 1.356 - 1.360
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.005 %
Free acid (as CH₃COOH) Max. 0.0005 %
Fluorescence (chinin) at 254 nm Max. 1.0 ppb
Fluorescence (chinin) at 365 nm Max. 1.0 ppb
Absorbance at 250 nm Max. 0.004
Absorbance at 230 nm Max. 0.02
Absorbance at 220 nm Max. 0.07
Absorbance at 210 nm Max. 0.20
Absorbance at 200 nm Max. 0.70
Explosion Limit ~8.3 %
Autoignition Temperature 500 °F

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty