Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acetonitrile ≥99.9% (by GC), CHROMASOLV™ for LC-MS, Riedel-de Haën™

Supplier: Honeywell Research Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: CyanomethaneEthanenitrileEthanoic acid nitrileMethane, cyano-Methyl cyanideMethanecarbonitrileACN

34967-2.5L 34967-4X2.5L 34967-4X4L 34967-1L 34967-250ML
BJ34967-2.5LEA 1340.61 USD InStock
BJ34967-2.5L BJ34967-250ML BJ34967-4X2.5L BJ34967-1L BJ34967-4L
Acetonitrile ≥99.9% (by GC), CHROMASOLV™ for LC-MS, Riedel-de Haën™
Acetonitrile


Formula: C₂H₃N
MW: 41.05 g/mol
Boiling Pt: 82 °C (1.013 hPa)
Melting Pt: -46 °C
Density: 0.7857 g/cm³ (20 °C) (lit.)
Flash Pt: 6 °C
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
Order Now
Specification Test Results
Assay (GC) Min. 99.9 %
Propionitrile (GC) Max. 0.2 %
Non-volatile matter Max. 0.0002 %
Water (Karl Fischer) Max. 0.01 %
Free acid (as CH₃COOH) Max. 0.001 %
Free alkali (as NH₃) Max. 0.0001 %
Silver (Ag) Max. 0.1 ppm
Aluminium (Al) Max. 0.5 ppm
Barium (Ba) Max. 0.1 ppm
Bismuth (Bi) Max. 0.1 ppm
Calcium (Ca) Max. 0.05 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.02 ppm
Chromium (Cr) Max. 0.02 ppm
Copper (Cu) Max. 0.02 ppm
Iron (Fe) Max. 0.02 ppm
Potassium (K) Max. 0.05 ppm
Lithium (Li) Max. 0.1 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.02 ppm
Molybdenum (Mo) Max. 0.1 ppm
Sodium (Na) Max. 0.05 ppm
Nickel (Ni) Max. 0.02 ppm
Lead (Pb) Max. 0.02 ppm
Tin (Sn) Max. 0.1 ppm
Strontium (Sr) Max. 0.1 ppm
Zinc (Zn) Max. 0.1 ppm
HPLC Gradient at 210 nm Max. 1 mAU
HPLC Gradient at 254 nm Max. 0.2 mAU
Baseline drift at 210 nm Max. 12 mAU
Suitability for the LC-MS complying
Fluorescence (chinin) at 254 nm Max. 0.5 ppb
Fluorescence (chinin) at 365 nm Max. 0.5 ppb
Absorbance at 195 nm Max. 0.097
Absorbance at 200 nm Max. 0.022
Absorbance at 230 nm Max. 0.004
Refractive Index n20/D 1.344 (lit.)

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty