Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

n-Heptane ≥99% (by GC), CHROMASOLV™ for LC-MS, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

34999-6X1L 34999-1L 34999-2.5L
BJ34999-1LCS 850.57 USD InStock
BJ34999-1L BJ-34999-2.5L BJ-34999-1L
n-Heptane ≥99% (by GC), CHROMASOLV™ for LC-MS, Riedel-de Haën™
n-Heptane
  • For LC-MS and LC-UV trace metals analysis
  • HPLC / UV-VIS quality control
  • Low gradient baseline drift
  • Low levels of trace metals
  • No non-volatile impurities


Formula: H₃C(CH₂)₅CH₃
MW: 100.20 g/mol
Boiling Pt: 98.2-98.4 °C (1.000 hPa)
Melting Pt: -90.5 °C
Density: 0.69 g/cm³ (15 °C)
Flash Pt: 6.7 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00009544
CAS Number: 142-82-5
EINECS: 205-563-8
UN: 1206
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/64/15/736415.jpg
Order Now
Specification Test Results
Assay (GC) min. 99 %
Non-volatile matter max. 0.0005 %
Water (Karl Fischer) max. 0.01 %
Free acid (as CH₃COOH) max. 0.001 %
Free alkali (as NH₃) max. 0.0005 %
Calcium (Ca) max. 0.00001 %
Potassium (K) max. 0.00001 %
Magnesium (Mg) max. 0.00001 %
Sodium (Na) max. 0.00001 %
Transmittance at 200 nm min. 20 %
Transmittance at 220 nm min. 80 %
Transmittance from 245 nm min. 98 %
Fluorescence (chinin) at 254 nm max. 1 ppb
Fluorescence (chinin) at 365 nm max. 1 ppb
Suitability for HPLC-MS complying

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty