Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 0.01 M HCl (0.01N) volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Synonyms: Muriatic acid

35334-6X1L 35334-2.5L
BJ35334-1LCS 655.62 USD InStock
BJ35334-1L BJ35334-2.5L
Hydrochloric acid 0.01 M HCl (0.01N) volumetric solution, Fluka™
Hydrochloric acid


Formula: HCl
MW: 36.46 g/mol
Boiling Pt: 100 °C (1.013 hPa)
Melting Pt: 0 °C
Density: 1.0 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-791-2
ADR: 8,III
Order Now
Specification Test Results
Factor 0.95 - 1.05
Ammonium (NH₄) Max. 3 ppm
Iron (Fe) Max. 0.1 ppm
Heavy metals (as Pb) Max. 0.5 ppm
Sulfate (SO₄) Max. 1 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty