Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Silver nitrate 0.1 M AgNO3 (0.1N) in aqueous solution ≥99% Reag. Ph. Eur. volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Warning
Synonyms: Silver mononitrateNitric acid silver saltLunar caustic

35375-5L-VP 35375-10L-VP 35375-6X1L 35375-2.5L 35375-500ML 35375-1L
BJ35375-5L-VPEA 736.97 USD InStock
BJ35375-5L-VP BJ35375-2.5L BJ35375-500ML BJ35375-1L BJ35375-10L-VP BJ35375-6X1L
Silver nitrate 0.1 M AgNO3 (0.1N) in aqueous solution ≥99% Reag. Ph. Eur. volumetric solution, Fluka™
Silver nitrate


Formula: AgNO₃
MW: 169.87 g/mol
Boiling Pt: 100 °C (1.013 hPa)
Melting Pt: <0 °C
Density: 1.010 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
UN: 3264
ADR: 8,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/45/16640645.jpg
Order Now
Specification Test Results
Factor 0.999 - 1.001
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective
Traceable to NIST SRM effective

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty