Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide ≥97%, beads 20-40 mesh, Reagent Grade

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Caustic sodaLyeSodium hydrateSoda lye

367176-2.5KG
BJ367176-2.5KGEA 146.59 USD InStock
BJ367176-2.5KG
Sodium hydroxide ≥97%, beads 20-40 mesh, Reagent Grade
Sodium hydroxide


Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1.390 °C (1.013 hPa)
Melting Pt: 319 °C
Density: 2.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II
Order Now
Specification Test Results
Form beads
Assay Min. 96.5 %
Carbonate (as Na₂CO₃) Max. 2.5 %
Granulation complying
Particle Size 20-40 mesh
Vapor Pressure <18 mmHg ( 20 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty