Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide concentrate for dilution to 10L, 1 M NaOH (1N) standard volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

38214-1EA
BJ38214-1EAEA 149.04 USD InStock
BJ38214-1EA
Sodium hydroxide concentrate for dilution to 10L, 1 M NaOH (1N) standard volumetric solution, Fluka™
Sodium hydroxide
concentrate for 10L standard solution


Formula: NaOH
MW: 40 g/mol
Boiling Pt: 120 °C
Melting Pt: 319 °C
Density: 1.350 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1824
ADR: 8,II
Order Now
Specification Test Results
Factor 0.998 - 1.002
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective
Traceable to NIST SRM effective

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty