Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chloroform ≥99.8% stabilized ACS, Riedel-de Haën™

Supplier: Honeywell Research Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: Methane, trichloro-TrichloroformTrichloromethane

Stabilization: Stabilised with amylene

472476-2.5L 472476-1L
BJ472476-2.5LEA 195.1 USD InStock
BJ472476-2.5L BJ472476-1L
Chloroform ≥99.8% stabilized ACS, Riedel-de Haën™
Chloroform
ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications


Formula: CHCl₃
MW: 119.38 g/mol
Boiling Pt: 61 °C (1.013 hPa)
Melting Pt: -61.3 °C
Density: 1.485 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/69/36/726936.jpg
Order Now
Specification Test Results
Identity (IR) complying
Assay (GC) Min. 99.80 %
2-methyl-2-buten (GC) 20 - 200 ppm
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.03 %
Free chlorine complying
Lead (Pb) Max. 0.05 ppm
Acetone, aldehydes (neßler test) complying
Free acid and chloride complying
Reaction against H₂SO₄ complying
APHA Max. 10
Contains amylenes as stabilizer

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty