Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Methyl-2-pyrrolidone (NMP) ≥99.7%, Biotechnology Grade, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: N-Methyl-alpha-pyrrolidone1-Methyl-2-pyrrolidoneNMPN-Methyl-2-pyrrolidoneN-Methylpyrrolidone1-Methyl-2-pyrrolidinoneN-MethylpyrrolidinoneN-methylpyrrolidoneN-Methyl-gamma-butyrolactamN-Methyl-2-pyrrolidinoneM-Pyrol

494496-4X2L 494496-2.5L 494496-2.5L 494496-1L 494496-1L
BJ494496-2.5LEA 806.39 USD InStock
BJ494496-2.5L BJ494496-1L BJ494496-2L CABJ494496-1L CABJ4944962.5L
N-Methyl-2-pyrrolidone (NMP) ≥99.7%, Biotechnology Grade, Burdick & Jackson™
N-Methyl-2-pyrrolidone (NMP)


Formula: C₅H₉NO
MW: 99.13 g/mol
Boiling Pt: 204 °C (1.013 hPa)
Melting Pt: -24 °C
Density: 1.028 g/cm³ (20 °C)
Flash Pt: 9.5 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00003193
CAS Number: 872-50-4
EINECS: 212-828-1
https://us-legacy.vwr.com/stibo/web/std.lang.all/88/54/2298854.jpg
Order Now
Specification Test Results
Assay (GC) Min. 99.7 %
Water (Karl Fischer) Max. 0.005 %
Titrable Base Max. 0.001 meq/g
Free alkali (as methylamine) Max. 0.01 %
Absorbance at 400 nm Max. 0.01
Absorbance at 350 nm Max. 0.01
Absorbance at 325 nm Max. 0.10
Absorbance at 300 nm Max. 0.50
Absorbance at 285 nm Max. 1.00
APHA Max. 20

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty