Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl acetate ≥99.8%, Biotechnology Grade, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Acetic acid ethyl esterAcetic etherAcetidinAcetoxyethaneEthyl acetic esterEthyl ethanoateVinegar naphtha

494518-2L 494518-1L 494518-1L 494518-2L
BJ494518-1LEA 144.58 USD InStock
CABJ494518-1L CABJ494518-2L BJ494518-1L BJ494518-2L
Ethyl acetate ≥99.8%, Biotechnology Grade, Burdick & Jackson™
Ethyl acetate


Formula: CH₃COOC₂H₅
MW: 88.11 g/mol
Boiling Pt: 77 °C (1.013 hPa)
Melting Pt: -84 °C
Density: 0.901 g/cm³
Flash Pt: 11.5 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/20/72/722072.jpg
Order Now
Specification Test Results
Identity (IR) complying
Assay (GC) Min. 99.8 %
Non-volatile matter Max. 0.0003 %
Water (Karl Fischer) Max. 0.01 %
Titrable Acid Max. 0.0009 meq/g
Absorbance at 400 nm Max. 0.01
Absorbance at 275 nm Max. 0.01
Absorbance at 263 nm Max. 0.05
Absorbance at 254 nm Max. 1.00

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty