Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acetonitrile ≥99.5% (by GC)

Supplier: Honeywell Research Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: Methane, cyano-Methyl cyanideEthanoic acid nitrileMethanecarbonitrileEthanenitrileCyanomethaneACN

60004-4X2.5L 60004-4X5L 60004-1L 60004-2.5L 60004-5L
BJ60004-4X2.5LCS 1441.99 USD InStock
BJ60004-4X2.5L BJ60004-1L BJ60004-2.5L BJ60004-5L BJ60004-4X5L
Acetonitrile ≥99.5% (by GC)
Acetonitrile
We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:

  • Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
  • Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.

Beilstein Number: 741857
RTECS Number: AL7700000
UNSPSC Code: 12191502


Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 82 °C (1.013 hPa)
Melting Pt: -46 °C
Density: 0.7857 g/cm³ (20 °C)
Flash Pt: 6 °C
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/89/10/718910.jpg
Order Now
Specification Test Results
Refractive Index n20/D 1.344 (lit.)
Assay (GC) Min. 99.5 %
Water (Karl Fischer) Max. 0.1 %
Vapor Density 1.41 (vs air)
Vapor Pressure 72.8 mmHg ( 20 °C)
Explosion Limit 16 %

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty