Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Chloroform ≥99.9% stabilized, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Methane, trichloro-TrichloroformTrichloromethane

Stabilization: Stabilised with amylene

650498-1L
BJ650498-1LEA 256.11 USD InStock
BJ650498-1L
Chloroform ≥99.9% stabilized, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Chloroform
contains amylenes as stabilizer

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities


Formula: CHCl₃
MW: 119.38 g/mol
Boiling Pt: 61 °C (1.013 hPa)
Melting Pt: -61.3 °C
Density: 1.485 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/69/36/726936.jpg
Order Now
Specification Test Results
Identity (IR) complying
Assay (GC) Min. 99.90 %
Non-volatile matter Max. 0.0001 %
Water (Karl Fischer) Max. 0.02 %
GC (as heptachlor epoxide) Max. 10 ng/l
2-methyl-2-buten (GC) 20 - 200 ppm
Absorbance at 400 nm Max. 0.005
Absorbance at 275 nm Max. 0.005
Absorbance at 250 nm Max. 0.30
Absorbance at 245 nm Max. 1.00
Fluorescence (chinin) at 365 nm Max. 0.5 ppb
Contains amylenes as stabilizer
Refractive Index n20/D 1.445 (lit.)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty